Assay Details
| Gene Symbol: POLDIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290145.1 | NP_001277074.1 | 1 - 2 | 303 | 101 | |
| NM_015584.4 | NP_056399.1 | 1 - 2 | 303 | 101 | ||
| GenBank mRNA | AF077203.1 | - | 1 - 2 | 211 | 101 | |
| AF179891.1 | - | 1 - 2 | 162 | 101 | ||
| AK313232.1 | - | 1 - 2 | 195 | 101 | ||
| BC000655.2 | - | 1 - 2 | 195 | 3349399 | 101 | |
| BC009265.2 | - | 1 - 2 | 189 | 3943941 | 101 | |
| BC018864.2 | - | 1 - 2 | 228 | 101 | ||
| BU932070.1 | - | 1 - 2 | 257 | 101 | ||
| DB455962.1 | - | 1 - 2 | 294 | 101 | ||
| HY130221.1 | - | 1 - 2 | 304 | 101 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001295.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17017098.hg.1 | ENST00000003607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017098.hg.1 | ENST00000540200.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017098.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017098.hg.1 | TC17001295.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017103.hg.1 | ENST00000003607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017103.hg.1 | ENST00000540200.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017103.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017103.hg.1 | TC17001295.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017104.hg.1 | ENST00000003607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017104.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017104.hg.1 | TC17001295.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017106.hg.1 | ENST00000003607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017106.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017106.hg.1 | TC17001295.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017107.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017107.hg.1 | TC17001295.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017099.hg.1 | ENST00000003607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017099.hg.1 | ENST00000540200.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017099.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17017099.hg.1 | TC17001295.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010191.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179682.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700179682.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700179682.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179682.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179682.hg.1 | TC1700010191.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700179682.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179682.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700179682.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700179682.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179686.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700179686.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179686.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179686.hg.1 | TC1700010191.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700179686.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700179686.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179686.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179687.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179687.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179687.hg.1 | TC1700010191.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700179687.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179687.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179681.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700179681.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700179681.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179681.hg.1 | NM_015584.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179681.hg.1 | TC1700010191.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700179681.hg.1 | ENST00000540200.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700179681.hg.1 | ENST00000618887.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700179681.hg.1 | NM_001290145.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700179681.hg.1 | NM_015584.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 142438 | - | - |

POLDIP2
Genomic Map