Assay Details
| Gene Symbol: PHF20L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016018.4 | NP_057102.4 | 6 - 7 | 832 | 104 | |
| XM_005250931.1 | XP_005250988.1 | 6 - 7 | 814 | 104 | ||
| XM_011517075.1 | XP_011515377.1 | 6 - 7 | 814 | 104 | ||
| XM_011517076.1 | XP_011515378.1 | 6 - 7 | 810 | 104 | ||
| XM_011517077.1 | XP_011515379.1 | 6 - 7 | 814 | 104 | ||
| XM_011517078.1 | XP_011515380.1 | 6 - 7 | 802 | 104 | ||
| XM_011517079.1 | XP_011515381.1 | 6 - 7 | 802 | 104 | ||
| XM_011517088.1 | XP_011515390.1 | 6 - 7 | 814 | 104 | ||
| XM_017013509.1 | XP_016868998.1 | 6 - 7 | 798 | 104 | ||
| XM_017013513.1 | XP_016869002.1 | 6 - 7 | 3637 | 104 | ||
| XM_017013514.1 | XP_016869003.1 | 6 - 7 | 3633 | 104 | ||
| XM_017013516.1 | XP_016869005.1 | 6 - 7 | 802 | 104 | ||
| GenBank mRNA | AB101203.1 | - | 6 - 7 | 799 | 104 | |
| AF151830.1 | - | 3 - 4 | 186 | 104 | ||
| BC002822.2 | - | 6 - 7 | 790 | 104 | ||
More Information
Set Membership: |
|

PHF20L1
Genomic Map