| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000222190.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000222190.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000222190.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000222190.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000222190.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000222190.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000598732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000598732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000598732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000598732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000598732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000598732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000600694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
ENST00000600694.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
NM_016145.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
NM_016145.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
NM_016145.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
NM_016145.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
NM_016145.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
NM_016145.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011890.hg.1 |
NM_016145.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19016883.hg.1 |
ENST00000222190.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19016883.hg.1 |
NM_016145.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19016883.hg.1 |
TC19001203.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000222190.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000222190.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000222190.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000222190.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000222190.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000222190.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000598732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000598732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000598732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000598732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000598732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000598732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000600694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
ENST00000600694.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
NM_016145.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
NM_016145.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
NM_016145.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
NM_016145.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
NM_016145.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
NM_016145.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011890.hg.1 |
NM_016145.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900204700.hg.1 |
ENST00000222190.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204700.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204700.hg.1 |
ENST00000598732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204700.hg.1 |
NM_016145.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900204700.hg.1 |
TC1900011890.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900204700.hg.1 |
ENST00000222190.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204700.hg.1 |
ENST00000596731.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204700.hg.1 |
ENST00000598732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900204700.hg.1 |
NM_016145.3 |
Refseq |