| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16018236.hg.1 |
ENST00000313732.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16018236.hg.1 |
ENST00000344861.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16018236.hg.1 |
NM_001145548.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16018236.hg.1 |
NM_017740.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16018236.hg.1 |
TC16001315.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1600186095.hg.1 |
ENST00000313732.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600186095.hg.1 |
ENST00000344861.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600186095.hg.1 |
ENST00000564466.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600186095.hg.1 |
ENST00000566909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600186095.hg.1 |
NM_001145548.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600186095.hg.1 |
NM_017740.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600186095.hg.1 |
TC1600011075.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600186095.hg.1 |
ENST00000313732.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600186095.hg.1 |
ENST00000344861.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600186095.hg.1 |
ENST00000564466.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600186095.hg.1 |
ENST00000566909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600186095.hg.1 |
NM_001145548.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600186095.hg.1 |
NM_017740.2 |
Refseq |