Assay Details
| Gene Symbol: MOB1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317112.1 | NP_001304041.1 | 1 - 2 | 310 | 75 | |
| NM_018221.4 | NP_060691.2 | 1 - 2 | 310 | 75 | ||
| GenBank mRNA | AB016839.1 | - | 1 - 2 | 159 | 75 | |
| AK001650.1 | - | 1 - 2 | 200 | 75 | ||
| AK021657.1 | - | 1 - 2 | 224 | 75 | ||
| BC003398.1 | - | 1 - 2 | 211 | 3448503 | 75 | |
| CN426611.1 | - | 1 - 2 | 198 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001993.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB016839.1,AK001650.1,AK021657.1,BC003398.1,NM_001317112.1,NM_018221.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02031740.hg.1 | ENST00000396049.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031740.hg.1 | ENST00000497054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031740.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031740.hg.1 | TC02001993.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031732.hg.1 | ENST00000396049.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031732.hg.1 | ENST00000409969.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031732.hg.1 | ENST00000497054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031732.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031732.hg.1 | TC02001993.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000463975.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000463975.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000463975.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000463975.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000463975.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000463975.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000488006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000488006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000488006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000494600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000494600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000494600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000495286.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000495286.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000495286.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000495286.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000495286.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000495286.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000495286.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000497054.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000497054.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000497054.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000497054.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000497054.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | ENST00000497054.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013096.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031731.hg.1 | ENST00000396049.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031731.hg.1 | ENST00000409969.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031731.hg.1 | ENST00000497054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031731.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031731.hg.1 | TC02001993.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031734.hg.1 | ENST00000396049.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031734.hg.1 | ENST00000409969.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031734.hg.1 | ENST00000497054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031734.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031734.hg.1 | TC02001993.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200195593.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195593.hg.1 | ENST00000463975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195593.hg.1 | ENST00000495286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195593.hg.1 | ENST00000497054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195593.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195593.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195593.hg.1 | ENST00000463975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195593.hg.1 | ENST00000495286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195593.hg.1 | ENST00000497054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195593.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195593.hg.1 | TC0200013096.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200195595.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195595.hg.1 | ENST00000463975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195595.hg.1 | ENST00000488006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195595.hg.1 | ENST00000495286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195595.hg.1 | ENST00000497054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195595.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195595.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195595.hg.1 | ENST00000463975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195595.hg.1 | ENST00000488006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195595.hg.1 | ENST00000495286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195595.hg.1 | ENST00000497054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195595.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195595.hg.1 | TC0200013096.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200195601.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195601.hg.1 | ENST00000463975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195601.hg.1 | ENST00000488006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195601.hg.1 | ENST00000494600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195601.hg.1 | ENST00000497054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195601.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195601.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195601.hg.1 | ENST00000463975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195601.hg.1 | ENST00000488006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195601.hg.1 | ENST00000494600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195601.hg.1 | ENST00000497054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195601.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195601.hg.1 | TC0200013096.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000463975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000463975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000463975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000463975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000463975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000463975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000488006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000488006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000488006.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000494600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000494600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000494600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000495286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000495286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000495286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000495286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000495286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000495286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000495286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000497054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000497054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000497054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000497054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000497054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | ENST00000497054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013096.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195594.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195594.hg.1 | ENST00000463975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195594.hg.1 | ENST00000495286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195594.hg.1 | ENST00000497054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195594.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195594.hg.1 | ENST00000396049.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195594.hg.1 | ENST00000463975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195594.hg.1 | ENST00000495286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195594.hg.1 | ENST00000497054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195594.hg.1 | NM_018221.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195594.hg.1 | TC0200013096.hg.1 | Clariom D TC |

MOB1A
Genomic Map