Assay Details
| Gene Symbol: PECR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018441.5 | NP_060911.2 | 5 - 6 | 678 | 62 | |
| XM_011511478.2 | XP_011509780.1 | 5 - 6 | 560 | 62 | ||
| GenBank mRNA | AF212234.1 | - | 5 - 6 | 663 | 62 | |
| AF232009.1 | - | 5 - 6 | 668 | 62 | ||
| AJ250303.1 | - | 5 - 6 | 664 | 62 | ||
| AK296208.1 | - | 4 - 5 | 570 | 62 | ||
| AK315795.1 | - | 5 - 6 | 637 | 62 | ||
| BC002529.2 | - | 5 - 6 | 642 | 3139219 | 62 | |
| BU151224.1 | - | 5 - 6 | 678 | 62 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02005052.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02044272.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044272.hg.1 | ENST00000497889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044272.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044272.hg.1 | TC02005052.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000464722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000464722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000464722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000464737.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000464737.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000464737.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000464737.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044275.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044275.hg.1 | ENST00000497889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044275.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02044275.hg.1 | TC02005052.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200228810.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228810.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228810.hg.1 | ENST00000464737.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228810.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228810.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200228810.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228810.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228810.hg.1 | ENST00000464737.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228810.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228810.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200228810.hg.1 | TC0200016768.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200228813.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228813.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228813.hg.1 | ENST00000464737.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228813.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228813.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200228813.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228813.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228813.hg.1 | ENST00000464737.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228813.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200228813.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200228813.hg.1 | TC0200016768.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000265322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000461330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000464722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000464722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000464722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000464737.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000464737.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000464737.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000464737.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000474093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000487318.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | ENST00000497889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016768.hg.1 | NM_018441.5 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 148928 | - | - |

PECR
Genomic Map