| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
ENST00000223864.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
ENST00000223864.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
ENST00000223864.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
ENST00000223864.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
ENST00000223864.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
ENST00000223864.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
ENST00000223864.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
ENST00000472145.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
ENST00000472145.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
ENST00000472145.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
ENST00000472145.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
ENST00000473877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
ENST00000473877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
ENST00000473877.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
ENST00000482696.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
ENST00000482696.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
ENST00000482696.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
ENST00000482696.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
ENST00000482696.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
NM_018465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
NM_018465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
NM_018465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
NM_018465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
NM_018465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
NM_018465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
NM_018465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009461.hg.1 |
NM_018465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011840.hg.1 |
ENST00000223864.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011840.hg.1 |
ENST00000482696.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011840.hg.1 |
NM_018465.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09011840.hg.1 |
TC09000891.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
ENST00000223864.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
ENST00000223864.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
ENST00000223864.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
ENST00000223864.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
ENST00000223864.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
ENST00000223864.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
ENST00000223864.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
ENST00000472145.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
ENST00000472145.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
ENST00000472145.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
ENST00000472145.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
ENST00000473877.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
ENST00000473877.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
ENST00000473877.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
ENST00000482696.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
ENST00000482696.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
ENST00000482696.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
ENST00000482696.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
ENST00000482696.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
NM_018465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
NM_018465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
NM_018465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
NM_018465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
NM_018465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
NM_018465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
NM_018465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009461.hg.1 |
NM_018465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900164946.hg.1 |
ENST00000223864.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900164946.hg.1 |
ENST00000472145.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900164946.hg.1 |
ENST00000482696.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900164946.hg.1 |
NM_018465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900164946.hg.1 |
ENST00000223864.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900164946.hg.1 |
ENST00000472145.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900164946.hg.1 |
ENST00000482696.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900164946.hg.1 |
NM_018465.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900164946.hg.1 |
TC0900009461.hg.1 |
Clariom D TC |