Assay Details
| Gene Symbol: S100A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020672.2 | NP_065723.1 | 2 - 3 | 147 | 92 | |
| XM_005245362.1 | XP_005245419.1 | 2 - 3 | 143 | 92 | ||
| XM_017001875.1 | XP_016857364.1 | 3 - 4 | 235 | 92 | ||
| GenBank mRNA | AY007220.1 | - | 2 - 3 | 129 | 92 | |
| BC005019.1 | - | 1 - 2 | 111 | 3640899 | 92 | |
| BG677973.1 | - | 2 - 3 | 147 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY007220.1, BC005019.1, NM_020672.2, XM_005245362.1, XM_017001875.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
response to lipopolysaccharide
toll-like receptor 4 signaling pathway
defense response to bacterium
calcium ion homeostasis
positive regulation of granulocyte chemotaxis
positive regulation of monocyte chemotaxis
calcium ion binding
protein binding
chemokine receptor binding
apoptotic process
response to lipopolysaccharide
toll-like receptor 4 signaling pathway
defense response to bacterium
calcium ion homeostasis
positive regulation of granulocyte chemotaxis
positive regulation of monocyte chemotaxis
calcium ion binding
protein binding
chemokine receptor binding
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00516782 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00516792 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00516776 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20574187 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00516777 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00516804 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00516780 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00516798 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00516807 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00516789 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00516781 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20574206 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20574178 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15130278 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00516775 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00516788 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00516786 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15130274 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15130276 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00516800 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00516795 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20574200 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20574174 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00516791 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20574208 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00516806 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00516803 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20574195 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20574196 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15130273 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00516801 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20574183 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20574199 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20574192 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 153131 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 61686 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 745513 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 956697 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P124619 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P124619 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 41813 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P107098 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P107098 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 56689 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P30310143 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_14_P107098 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P30310143 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P124619 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P124619 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516792 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516803 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516781 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20574206 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15130276 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20574192 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20574208 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516780 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20574196 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516776 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516777 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20574183 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20574200 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20574178 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516807 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516801 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516806 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20574195 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516775 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516791 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516789 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516788 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516786 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20574174 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516795 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20574187 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20574183 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516789 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516788 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516791 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516776 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20574195 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516803 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516792 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20574187 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516781 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20574178 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20574208 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516801 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516795 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20574192 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516780 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516806 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516807 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516786 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20574206 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20574200 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20574174 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516775 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15130276 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00516777 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20574196 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P124619 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P107098 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00516776 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00516798 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00516783 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00516804 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00516779 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00516801 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00516805 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00516774 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00516785 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00516807 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00516803 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00516794 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00516789 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00516780 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00516787 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00516806 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00516792 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P107098 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P107098 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P124619 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 58836 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P107098 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516801 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516758 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516744 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516787 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516749 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516765 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15130277 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516772 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516737 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516755 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516794 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516729 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516806 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516785 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516798 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516789 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516718 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516713 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516708 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516728 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516719 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516768 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516725 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15130276 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516715 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516819 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516756 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516770 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516774 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516735 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516732 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516721 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516741 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516760 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516711 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516747 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516779 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516762 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516742 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516776 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516706 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516783 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516792 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516751 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516733 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516740 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516780 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00516753 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15130271 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15130273 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P107098 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_020672_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7920300 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7920299 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7920298 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7920301 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7920297 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 218677_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11720803_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11720803_at | - | - |
| Human Genome U133 Plus 2.0 Array | 218677_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 218677_at | - | - |
| Human Genome U133A 2.0 Array | 218677_at | - | - |
| Human Genome U133A 2.0 Array | 218677_PM_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_020672_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw516457 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 218677_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 218677_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 218677_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 218677_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 218677_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 218677_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 218677_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 218677_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 218677_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 218677_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 218677_at8 | - | - |
| Human Genome U95 Set | 89770_at | - | - |
| Human Genome U95 Set | 89770_PM_at | - | - |
| Human Genome U133 Set | 218677_at | - | - |
| Human Genome U133 Set | 218677_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A052350 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A052350 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 185644 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1783287 | NM_020672.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1783287 | AY007220.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1783287 | NM_020672.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1783287 | AY007220.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1783287 | NM_020672.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1783287 | AY007220.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1783287 | NM_020672.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1783287 | AY007220.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1783287 | NM_020672.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1783287 | AY007220.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_10190711-S | NM_020672.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_10190711-S | AY007220.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_17182-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0002900114 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0002900114 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1783287 | NM_020672.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1783287 | AY007220.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1783287 | NM_020672.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1783287 | AY007220.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 2470039 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_17182 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1783287 | NM_020672.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1783287 | AY007220.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1783287 | NM_020672.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1783287 | AY007220.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1783287 | NM_020672.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1783287 | AY007220.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1783287 | NM_020672.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1783287 | AY007220.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1783287 | NM_020672.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1783287 | AY007220.1 | human_transcriptome |

S100A14
Genomic Map