Assay Details
| Gene Symbol: ZFP69B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005271138.3 | XP_005271195.1 | 2 - 3 | 538 | 69 | |
| XM_005271139.3 | XP_005271196.1 | 2 - 3 | 538 | 69 | ||
| GenBank mRNA | AL137241.1 | - | 2 - 3 | 490 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AL137241.1, XM_005271138.3, XM_005271139.3 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000361584.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000411995.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000469416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | ENST00000484445.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | NM_023070.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007958.hg.1 | NM_023070.2 | Refseq |

ZFP69B
Genomic Map