Assay Details
| Gene Symbol: PCYOX1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301054.1 | NP_001287983.1 | 1 - 2 | 170 | 90 | |
| NM_001301057.1 | NP_001287986.1 | 1 - 2 | 170 | 90 | ||
| GenBank mRNA | AF451985.1 | - | 1 - 2 | 115 | 90 | |
| BC000014.2 | - | 1 - 2 | 106 | 3506623 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000274569.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000274569.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000274569.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000274569.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000274569.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000274569.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000274569.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000274569.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000274569.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000274569.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000274569.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000505669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000505669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000505669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000505669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000505669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000505669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000505669.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000507621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000507621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000507621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000507621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000507621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000507621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000507621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000507621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000507621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000510990.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000510990.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000510990.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000510990.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000510990.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000510990.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000511945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000511945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000511945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000511945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000511945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000511945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000511945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000511945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000511945.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000514349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000514349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000514349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000514349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000514349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000514349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000514349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000514349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | ENST00000514349.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_001301054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_001301054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_001301054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_001301054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_001301054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_001301054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_001301054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_001301054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_001301054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_001301054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_001301054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_001301057.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_001301057.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_001301057.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_001301057.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_001301057.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_001301057.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_001301057.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_001301057.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_001301057.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_024028.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_024028.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_024028.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_024028.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_024028.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_024028.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_024028.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_024028.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_024028.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_024028.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009061.hg.1 | NM_024028.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000274569.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000274569.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000274569.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000274569.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000274569.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000274569.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000274569.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000274569.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000274569.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000274569.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000274569.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000505669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000505669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000505669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000505669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000505669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000505669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000505669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000507621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000507621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000507621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000507621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000507621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000507621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000507621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000507621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000507621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000510990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000510990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000510990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000510990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000510990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000510990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000511945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000511945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000511945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000511945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000511945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000511945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000511945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000511945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000511945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000514349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000514349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000514349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000514349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000514349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000514349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000514349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000514349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | ENST00000514349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_001301054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_001301054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_001301054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_001301054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_001301054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_001301054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_001301054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_001301054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_001301054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_001301054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_001301054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_001301057.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_001301057.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_001301057.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_001301057.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_001301057.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_001301057.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_001301057.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_001301057.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_001301057.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_024028.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_024028.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_024028.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_024028.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_024028.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_024028.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_024028.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_024028.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_024028.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_024028.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009061.hg.1 | NM_024028.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163177.hg.1 | ENST00000274569.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163177.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163177.hg.1 | ENST00000505669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163177.hg.1 | ENST00000510990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163177.hg.1 | ENST00000511945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163177.hg.1 | ENST00000514349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163177.hg.1 | NM_001301054.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163177.hg.1 | NM_001301057.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163177.hg.1 | NM_024028.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163177.hg.1 | ENST00000274569.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163177.hg.1 | ENST00000503240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163177.hg.1 | ENST00000505669.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163177.hg.1 | ENST00000510990.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163177.hg.1 | ENST00000511945.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163177.hg.1 | ENST00000514349.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500163177.hg.1 | NM_001301054.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163177.hg.1 | NM_001301057.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163177.hg.1 | NM_024028.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500163177.hg.1 | TC0500009061.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 166964 | - | - |

PCYOX1L
Genomic Map