Assay Details
| Gene Symbol: DBF4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011525281.2 | XP_011523583.1 | 4 - 5 | 499 | 89 | |
| XM_017025152.1 | XP_016880641.1 | 4 - 5 | 499 | 89 | ||
| XM_017025153.1 | XP_016880642.1 | 4 - 5 | 494 | 89 | ||
| GenBank mRNA | AK023149.1 | - | 4 - 5 | 583 | 89 | |
| AK295296.1 | - | 5 - 6 | 476 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011525281.2, XM_017025152.1, XM_017025153.1 transcripts.
Set Membership: |
|

DBF4B
Genomic Map