Assay Details
| Gene Symbol: FLAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184892.1 | NP_001171821.1 | 2 - 3 | 328 | 69 | |
| GenBank mRNA | AF218022.1 | - | 2 - 3 | 255 | 69 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001282.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF218022.1,NM_001184892.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000292180.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000295530.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000315144.14 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368431.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000368433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000477609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000477609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000477609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000477609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000477609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000481758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000481758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000481758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000481758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000481758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000487371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000487371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000487371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000487371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000487371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000489992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000489992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000489992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000489992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000489992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000489992.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000492620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000492620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000492620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | ENST00000492620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_001184892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_025207.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010102.hg.1 | NM_201398.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 118224 | - | - |

FLAD1
Genomic Map