Assay Details
| Gene Symbol: TMUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136044.1 | NP_001129516.1 | 1 - 2 | 100 | 107 | |
| GenBank mRNA | AY037155.1 | - | 1 - 2 | 81 | 109 | |
| BC000936.1 | - | 1 - 2 | 100 | 3449979 | 107 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07002030.hg.1
Additional Information:
This assay is in the 5'UTR region of the AY037155.1,BC000936.1,NM_001136044.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07031061.hg.1 | ENST00000297533.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07031061.hg.1 | ENST00000392818.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07031061.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07031061.hg.1 | ENST00000476627.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07031061.hg.1 | ENST00000482202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07031061.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07031061.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07031061.hg.1 | TC07002030.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000488752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000488752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000488752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000488752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000488752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000492838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000492838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000492838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | ENST00000492838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07031075.hg.1 | ENST00000297533.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07031075.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07031075.hg.1 | TC07002030.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0700194177.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194177.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194177.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194177.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194177.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194177.hg.1 | ENST00000488752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194177.hg.1 | ENST00000492838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194177.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700194177.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700194177.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194177.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194177.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194177.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194177.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194177.hg.1 | ENST00000488752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194177.hg.1 | ENST00000492838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194177.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700194177.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700194177.hg.1 | TC0700013060.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700194178.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194178.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194178.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194178.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194178.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194178.hg.1 | ENST00000488752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194178.hg.1 | ENST00000492838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194178.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700194178.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700194178.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194178.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194178.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194178.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194178.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194178.hg.1 | ENST00000488752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194178.hg.1 | ENST00000492838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194178.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700194178.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700194178.hg.1 | TC0700013060.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700194191.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194191.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700194191.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194191.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700194191.hg.1 | TC0700013060.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000297533.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000392818.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000462940.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000476627.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000482202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000488752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000488752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000488752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000488752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000488752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000492838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000492838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000492838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | ENST00000492838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_001136044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013060.hg.1 | NM_031434.3 | Refseq |

TMUB1
Genomic Map