Assay Details
| Gene Symbol: MMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145938.1 | NP_001139410.1 | 6 - 7 | 892 | 133 | |
| NM_002421.3 | NP_002412.1 | 6 - 7 | 1044 | 133 | ||
| GenBank mRNA | AK024818.1 | - | 6 - 7 | 967 | 133 | |
| AK098450.1 | - | 7 - 8 | 899 | 133 | ||
| AK130687.1 | - | 6 - 7 | 967 | 133 | ||
| AK223034.1 | - | 5 - 6 | 670 | 133 | ||
| AK223035.1 | - | 6 - 7 | 925 | 133 | ||
| AK223036.1 | - | 4 - 5 | 508 | 133 | ||
| AK223056.1 | - | 5 - 6 | 710 | 133 | ||
| AK223281.1 | - | 6 - 7 | 967 | 133 | ||
| AK225495.1 | - | 5 - 6 | 735 | 133 | ||
| AK297723.1 | - | 6 - 7 | 815 | 133 | ||
| AK301340.1 | - | 6 - 7 | 757 | 133 | ||
| BC013118.1 | - | 2 - 3 | 192 | 133 | ||
| BC013875.2 | - | 6 - 7 | 943 | 3834572 | 133 | |
| BT006874.1 | - | 6 - 7 | 901 | 133 | ||
| BT020147.1 | - | 6 - 7 | 901 | 133 | ||
| M13509.1 | - | 6 - 7 | 969 | 133 | ||
| X05231.1 | - | 6 - 7 | 967 | 133 | ||
| X54925.1 | - | 6 - 7 | 969 | 133 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11002234.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (23)
Up-regulation of alveolar macrophage matrix metalloproteinases in HIV1(+) smokers with early emphysema.
- Authors
- Kaner RJ, Santiago F, Crystal RG
- Journal
- J Leukoc Biol - Page No 913 (Thu Oct 01 00:00:00 UTC 2009)
- Volume (Year)
- 86 (2009)
- Product Usage
- -
Role of hypoxia-inducible factor-1alpha in hypoxia-induced expressions of IL-8, MMP-1 and MMP-3 in rheumatoid fibroblast-like synoviocytes.
- Authors
- Ahn JK, Koh EM, Cha HS, Lee YS, Kim J, Bae EK, Ahn KS
- Journal
- Rheumatology (Oxford) - Page No 834 (Sun Jun 01 00:00:00 UTC 2008)
- Volume (Year)
- 47 (2008)
- Product Usage
- -
Effects of 10 cigarette smoke condensates on primary human airway epithelial cells by comparative gene and cytokine expression studies.
- Authors
- Pickett G, Seagrave J, Boggs S, Polzin G, Richter P, Tesfaigzi Y
- Journal
- Toxicol Sci - Page No 79 (Mon Mar 01 00:00:00 UTC 2010)
- Volume (Year)
- 114 (2010)
- Product Usage
- -
Mechanisms of the inhibitory effect of epigallocatechin-3-gallate on cultured human vascular smooth muscle cell invasion.
- Authors
- Cheng XW, Kuzuya M, Nakamura K, Liu Z, Di Q, Hasegawa J, Iwata M, Murohara T, Yokota M, Iguchi A
- Journal
- Arterioscler Thromb Vasc Biol - Page No 1864 (Thu Sep 01 00:00:00 UTC 2005)
- Volume (Year)
- 25 (2005)
- Product Usage
- -
Identification of molecular distinctions between normal breast-associated fibroblasts and breast cancer-associated fibroblasts.
- Authors
- Sadlonova A, Bowe DB, Novak Z, Mukherjee S, Duncan VE, Page GP, Frost AR
- Journal
- Cancer Microenviron - Page No 9 (Tue Dec 01 00:00:00 UTC 2009)
- Volume (Year)
- 2 (2009)
- Product Usage
- -
Matrix and TGF-beta-related gene expression during human dental pulp stem cell (DPSC) mineralization.
- Authors
- Liu J, Jin T, Chang S, Ritchie HH, Smith AJ, Clarkson BH
- Journal
- In Vitro Cell Dev Biol Anim - Page No 120 (Thu Mar 01 00:00:00 UTC 2007)
- Volume (Year)
- 43 (2007)
- Product Usage
- -
Phagocytosis and remodeling of collagen matrices.
- Authors
- Abraham LC, Dice JF, Lee K, Kaplan DL
- Journal
- Exp Cell Res - Page No 1045 (Sat Mar 10 00:00:00 UTC 2007)
- Volume (Year)
- 313 (2007)
- Product Usage
- -
The expression of proinflammatory cytokines and matrix metalloproteinases in the synovial membranes of patients with osteoarthritis compared with traumatic knee disorders.
- Authors
- Wassilew GI, Lehnigk U, Duda GN, Taylor WR, Matziolis G, Dynybil C
- Journal
- Arthroscopy - Page No 1096 (Sun Aug 01 00:00:00 UTC 2010)
- Volume (Year)
- 26 (2010)
- Product Usage
- -
Relation between intracavernosal dose of prostaglandin Pge 1 and mean duration of erection in men with different underlying causes of erectile dysfunction.
- Authors
- Bratus D, Hlebic G, Hajdinjak T
- Journal
- Croat Med J - Page No 76 (Thu Feb 01 00:00:00 UTC 2007)
- Volume (Year)
- 48 (2007)
- Product Usage
- -
UV-induced DNA damage initiates release of MMP-1 in human skin.
- Authors
- Dong KK, Damaghi N, Picart SD, Markova NG, Obayashi K, Okano Y, Masaki H, Grether-Beck S, Krutmann J, Smiles KA, Yarosh DB
- Journal
- Exp Dermatol - Page No 1037 (Mon Dec 01 00:00:00 UTC 2008)
- Volume (Year)
- 17 (2008)
- Product Usage
- -
Protein isoprenylation regulates secretion of matrix metalloproteinase 1 from rheumatoid synovial fibroblasts: effects of statins and farnesyl and geranylgeranyl transferase inhibitors.
- Authors
- Abeles AM, Marjanovic N, Park J, Attur M, Chan ES, Al-Mussawir HE, Dave M, Fisher MC, Stuchin SA, Abramson SB, Pillinger MH
- Journal
- Arthritis Rheum - Page No 2840 (Sat Sep 01 00:00:00 UTC 2007)
- Volume (Year)
- 56 (2007)
- Product Usage
- -
Impact of collagen structure on matrix trafficking by human fibroblasts.
- Authors
- Abraham LC, Vorrasi J, Kaplan DL
- Journal
- J Biomed Mater Res A - Page No 39 (Thu Jul 01 00:00:00 UTC 2004)
- Volume (Year)
- 70 (2004)
- Product Usage
- -
Liver X receptor is a therapeutic target for photoaging and chronological skin aging.
- Authors
- Chang KC, Shen Q, Oh IG, Jelinsky SA, Jenkins SF, Wang W, Wang Y, LaCava M, Yudt MR, Thompson CC, Freedman LP, Chung JH, Nagpal S
- Journal
- Mol Endocrinol - Page No 2407 (Sat Nov 01 00:00:00 UTC 2008)
- Volume (Year)
- 22 (2008)
- Product Usage
- -
Anabolic and catabolic responses of human articular chondrocytes to varying oxygen percentages.
- Authors
- Strobel S, Loparic M, Wendt D, Schenk AD, Candrian C, Lindberg RL, Moldovan F, Barbero A, Martin I
- Journal
- Arthritis Res Ther - Page No R34 (Fri Jan 01 00:00:00 UTC 2010)
- Volume (Year)
- 12 (2010)
- Product Usage
- -
Altered production of extra-cellular matrix components by muscle-derived Duchenne muscular dystrophy fibroblasts before and after TGF-beta1 treatment.
- Authors
- Zanotti S, Gibertini S, Mora M
- Journal
- Cell Tissue Res - Page No 397 (Mon Feb 01 00:00:00 UTC 2010)
- Volume (Year)
- 339 (2010)
- Product Usage
- -
First look at the effect of overexpression of TIGR/MYOC on the transcriptome of the human trabecular meshwork.
- Authors
- Borras T, Bryant PA, Chisolm SS
- Journal
- Exp Eye Res - Page No 1002 (Thu Jun 01 00:00:00 UTC 2006)
- Volume (Year)
- 82 (2006)
- Product Usage
- -
Nitric oxide donors selectively reduce the expression of matrix metalloproteinases-8 and -9 by human diabetic skin fibroblasts.
- Authors
- Burrow JW, Koch JA, Chuang HH, Zhong W, Dean DD, Sylvia VL
- Journal
- J Surg Res - Page No 90 (Fri Jun 01 00:00:00 UTC 2007)
- Volume (Year)
- 140 (2007)
- Product Usage
- -
Cervical tissue engineering using silk scaffolds and human cervical cells.
- Authors
- House M, Sanchez CC, Rice WL, Socrate S, Kaplan DL
- Journal
- Tissue Eng Part A - Page No 2101 (Tue Jun 01 00:00:00 UTC 2010)
- Volume (Year)
- 16 (2010)
- Product Usage
- -
Induction of matrix metalloproteinase-1 by small interfering RNA targeting connective tissue growth factor in dermal fibroblasts from patients with systemic sclerosis.
- Authors
- Ishibuchi H, Abe M, Yokoyama Y, Ishikawa O
- Journal
- Exp Dermatol - Page No e111 (Sun Aug 01 00:00:00 UTC 2010)
- Volume (Year)
- 19 (2010)
- Product Usage
- -
Signal transducer and activator of transcription 3 activation promotes invasive growth of colon carcinomas through matrix metalloproteinase induction.
- Authors
- Tsareva SA, Moriggl R, Corvinus FM, Wiederanders B, Schutz A, Kovacic B, Friedrich K
- Journal
- Neoplasia - Page No 279 (Sun Apr 01 00:00:00 UTC 2007)
- Volume (Year)
- 9 (2007)
- Product Usage
- -
Multiplex preamplification of specific cDNA targets prior to gene expression analysis by TaqMan Arrays.
- Authors
- Mengual L, Burset M, Marin-Aguilera M, Ribal MJ, Alcaraz A
- Journal
- Bmc Res Notes - Page No 21 (Tue Jan 01 00:00:00 UTC 2008)
- Volume (Year)
- 1 (2008)
- Product Usage
- -
Role for cathepsin K in emphysema in smoke-exposed guinea pigs.
- Authors
- Golovatch P, Mercer BA, Lemaitre V, Wallace A, Foronjy RF, D'Armiento J
- Journal
- Exp Lung Res - Page No 631 (Thu Oct 01 00:00:00 UTC 2009)
- Volume (Year)
- 35 (2009)
- Product Usage
- -
Adult human bone marrow stromal cells regulate expression of their MMPs and TIMPs in differentiation type-specific manner.
- Authors
- Mauney J, Volloch V
- Journal
- Matrix Biol - Page No 3 (Fri Jan 01 00:00:00 UTC 2010)
- Volume (Year)
- 29 (2010)
- Product Usage
- -

MMP1
Genomic Map