Assay Details
| Gene Symbol: FDXR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004110.5 | NP_004101.3 | 6 - 7 | 700 | 65 | |
| XM_011524527.1 | XP_011522829.1 | 6 - 7 | 648 | 65 | ||
| XM_011524528.1 | XP_011522830.1 | 6 - 7 | 737 | 65 | ||
| XM_011524529.1 | XP_011522831.1 | 6 - 7 | 740 | 65 | ||
| XM_011524531.2 | XP_011522833.1 | 5 - 6 | 633 | 65 | ||
| XM_011524532.2 | XP_011522834.1 | 5 - 6 | 640 | 65 | ||
| GenBank mRNA | J03826.1 | - | 6 - 7 | 633 | 65 | |
More Information
Set Membership: |
|

FDXR
Genomic Map