Assay Details
| Gene Symbol: DSC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004949.4 | NP_004940.1 | 1 - 2 | 531 | 111 | |
| NM_024422.4 | NP_077740.1 | 1 - 2 | 531 | 111 | ||
| GenBank mRNA | AK290313.1 | - | 1 - 2 | 222 | 111 | |
| AK297029.1 | - | 1 - 2 | 158 | 111 | ||
| BC063291.1 | - | 1 - 2 | 513 | 30337780 | 111 | |
| DC382631.1 | - | 1 - 2 | 531 | 111 | ||
| X56807.1 | - | 1 - 2 | 110 | 111 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000436.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK290313.1,AK297029.1,BC063291.1,NM_004949.4,NM_024422.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
cellular response to starvation
cardiac muscle cell-cardiac muscle cell adhesion
bundle of His cell-Purkinje myocyte adhesion involved in cell communication
regulation of heart rate by cardiac conduction
regulation of ventricular cardiac muscle cell action potential
calcium ion binding
protein binding
cell adhesive protein binding involved in bundle of His cell-Purkinje myocyte communication
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
cellular response to starvation
cardiac muscle cell-cardiac muscle cell adhesion
bundle of His cell-Purkinje myocyte adhesion involved in cell communication
regulation of heart rate by cardiac conduction
regulation of ventricular cardiac muscle cell action potential
calcium ion binding
protein binding
cell adhesive protein binding involved in bundle of His cell-Purkinje myocyte communication
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR18004454.hg.1 | ENST00000280904.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004454.hg.1 | ENST00000399347.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004454.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004454.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004454.hg.1 | TC18000436.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004451.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004451.hg.1 | ENST00000280904.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004451.hg.1 | ENST00000399347.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004451.hg.1 | ENST00000438199.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004451.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004451.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004451.hg.1 | TC18000436.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004453.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004453.hg.1 | ENST00000280904.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004453.hg.1 | ENST00000399347.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004453.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004453.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004453.hg.1 | TC18000436.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004455.hg.1 | ENST00000280904.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004455.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004455.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004455.hg.1 | TC18000436.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1800163959.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163959.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800163959.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800163959.hg.1 | TC1800009268.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800163959.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163959.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800163959.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800163962.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800163962.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800163962.hg.1 | TC1800009268.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800163962.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800163962.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800163960.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163960.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800163960.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800163960.hg.1 | TC1800009268.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800163960.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163960.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800163960.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800163961.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163961.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800163961.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800163961.hg.1 | TC1800009268.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800163961.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163961.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800163961.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | ENST00000251081.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_004949.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_024422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009268.hg.1 | NM_024422.4 | Refseq |

DSC2
Genomic Map