Assay Details
| Gene Symbol: CCR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006641.3 | NP_006632.2 | 1 - 2 | 153 | 65 | |
| GenBank mRNA | AF145440.1 | - | 1 - 2 | 98 | 65 | |
| DB132476.1 | - | 1 - 2 | 148 | 65 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000251.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF145440.1,NM_006641.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000355983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000355983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000355983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000355983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000355983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000355983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000355983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000355983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000357632.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000357632.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000357632.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000357632.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000357632.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000357632.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000357632.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000395963.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000395963.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000395963.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000395963.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000395963.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000395963.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000422395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000422395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000422395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000422395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000422395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000463197.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000463197.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000463197.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | ENST00000463197.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | NM_001256369.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | NM_001256369.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | NM_001256369.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | NM_001256369.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | NM_001256369.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | NM_001256369.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | NM_001256369.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | NM_001256369.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | NM_006641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | NM_006641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | NM_006641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | NM_006641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | NM_006641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | NM_006641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | NM_031200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | NM_031200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | NM_031200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | NM_031200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | NM_031200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | NM_031200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007247.hg.1 | NM_031200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005132.hg.1 | ENST00000355983.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005132.hg.1 | ENST00000357632.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005132.hg.1 | ENST00000395963.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005132.hg.1 | ENST00000422395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005132.hg.1 | NM_001256369.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005132.hg.1 | NM_006641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005132.hg.1 | NM_031200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005132.hg.1 | TC03000251.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005133.hg.1 | ENST00000357632.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005133.hg.1 | ENST00000395963.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005133.hg.1 | NM_001256369.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005133.hg.1 | NM_006641.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005133.hg.1 | NM_031200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005133.hg.1 | TC03000251.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0300151318.hg.1 | ENST00000355983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151318.hg.1 | ENST00000357632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151318.hg.1 | ENST00000395963.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151318.hg.1 | ENST00000422395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151318.hg.1 | NM_001256369.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151318.hg.1 | NM_006641.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151318.hg.1 | NM_031200.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151318.hg.1 | ENST00000355983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151318.hg.1 | ENST00000357632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151318.hg.1 | ENST00000395963.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151318.hg.1 | ENST00000422395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151318.hg.1 | NM_001256369.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151318.hg.1 | NM_006641.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151318.hg.1 | NM_031200.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151318.hg.1 | TC0300007247.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300151320.hg.1 | ENST00000355983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151320.hg.1 | ENST00000357632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151320.hg.1 | ENST00000395963.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151320.hg.1 | NM_001256369.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151320.hg.1 | NM_006641.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151320.hg.1 | NM_031200.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151320.hg.1 | ENST00000355983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151320.hg.1 | ENST00000357632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151320.hg.1 | ENST00000395963.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151320.hg.1 | NM_001256369.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151320.hg.1 | NM_006641.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151320.hg.1 | NM_031200.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151320.hg.1 | TC0300007247.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000355983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000355983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000355983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000355983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000355983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000355983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000355983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000355983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000357632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000357632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000357632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000357632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000357632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000357632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000357632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000395963.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000395963.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000395963.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000395963.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000395963.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000395963.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000422395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000422395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000422395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000422395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000422395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000463197.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000463197.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000463197.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | ENST00000463197.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | NM_001256369.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | NM_001256369.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | NM_001256369.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | NM_001256369.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | NM_001256369.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | NM_001256369.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | NM_001256369.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | NM_001256369.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | NM_006641.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | NM_006641.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | NM_006641.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | NM_006641.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | NM_006641.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | NM_006641.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | NM_031200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | NM_031200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | NM_031200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | NM_031200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | NM_031200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | NM_031200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007247.hg.1 | NM_031200.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151319.hg.1 | ENST00000355983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151319.hg.1 | ENST00000357632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151319.hg.1 | ENST00000395963.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151319.hg.1 | ENST00000422395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151319.hg.1 | NM_001256369.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151319.hg.1 | NM_006641.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151319.hg.1 | NM_031200.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151319.hg.1 | ENST00000355983.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151319.hg.1 | ENST00000357632.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151319.hg.1 | ENST00000395963.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151319.hg.1 | ENST00000422395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300151319.hg.1 | NM_001256369.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151319.hg.1 | NM_006641.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151319.hg.1 | NM_031200.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300151319.hg.1 | TC0300007247.hg.1 | Clariom D TC |

CCR9
Genomic Map