Assay Details
| Gene Symbol: F8A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012151.3 | NP_036283.2 | 1 | 177 | 66 | |
| GenBank mRNA | AL556125.3 | - | 1 | 175 | 66 | |
| BC039693.1 | - | 1 | 163 | 5214628 | 66 | |
| BC071963.1 | - | 1 | 167 | 5477018 | 66 | |
| Gene Symbol: F8A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007523.1 | NP_001007524.1 | 1 | 162 | 66 | |
| Gene Symbol: F8A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007524.1 | NP_001007525.1 | 1 | 162 | 66 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000781.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3242234 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 47801 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3242234 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3242234 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3242234 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8176247 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8171000 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8170999 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8171031 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8176246 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8170999 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8171030 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8176247 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8171030 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8171029 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8176245 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8170998 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8176245 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8171029 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8170998 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008874.hg.1 | ENST00000369505.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X010609.hg.1 | ENST00000369446.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X010609.hg.1 | NM_001007523.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X010609.hg.1 | NM_001007524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X010609.hg.1 | NM_012151.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X010609.hg.1 | TC0X000781.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011244.hg.1 | ENST00000622749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011244.hg.1 | ENST00000622749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011244.hg.1 | ENST00000622749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008860.hg.1 | ENST00000610495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008860.hg.1 | ENST00000610495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008860.hg.1 | NM_001007523.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008860.hg.1 | NM_001007524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008860.hg.1 | NM_012151.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008860.hg.1 | NM_012151.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00160779.hg.1 | ENST00000610495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00160779.hg.1 | NM_001007523.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00160779.hg.1 | NM_001007524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00160779.hg.1 | NM_012151.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00160779.hg.1 | ENST00000610495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00160779.hg.1 | NM_001007523.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00160779.hg.1 | NM_001007524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00160779.hg.1 | NM_012151.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00160779.hg.1 | TC0X00008860.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00008874.hg.1 | ENST00000369505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011244.hg.1 | ENST00000622749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011244.hg.1 | ENST00000622749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011244.hg.1 | ENST00000622749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008860.hg.1 | ENST00000610495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008860.hg.1 | ENST00000610495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008860.hg.1 | NM_001007523.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008860.hg.1 | NM_001007524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008860.hg.1 | NM_012151.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008860.hg.1 | NM_012151.3 | Refseq |
| Human Genome U133 Plus 2.0 Array | 203274_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 203274_at | - | - |
| Human Genome U133 Plus 2.0 Array | 203274_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 203274_at | - | - |
| Human Genome U133A 2.0 Array | 203274_PM_at | - | - |
| Human Genome U133A 2.0 Array | 203274_at | - | - |
| Human Genome U133A 2.0 Array | 203274_at | - | - |
| Human Genome U133A 2.0 Array | 203274_PM_at | - | - |
| Human Genome U95 Set | 37907_at | - | - |
| Human Genome U95 Set | 37907_PM_at | - | - |
| Human Genome U95 Set | 37907_PM_at | - | - |
| Human Genome U95 Set | 37907_at | - | - |
| Human Genome U133 Set | 203274_at | - | - |
| Human Genome U133 Set | 203274_at | - | - |
| Human Genome U133 Set | 203274_PM_at | - | - |
| Human Genome U133 Set | 203274_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A041788 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A041788 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A041788 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A041788 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1748092 | NM_001007523.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1748092 | NM_001007524.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1748092 | CR623398.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1748092 | CR624283.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1748092 | CR621827.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1727984 | NM_001007523.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1727984 | NM_001007524.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1727984 | CR623398.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1727984 | CR624283.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1727984 | CR621827.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3247893 | NM_001007523.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3247893 | NM_001007524.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3247893 | CR623398.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3247893 | CR624283.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3247893 | CR621827.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1727984 | NM_001007523.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1727984 | NM_001007524.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1727984 | CR623398.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1727984 | CR624283.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1727984 | CR621827.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1727984 | NM_001007523.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1727984 | NM_001007524.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1727984 | CR623398.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1727984 | CR624283.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1727984 | CR621827.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3247893 | NM_001007523.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3247893 | NM_001007524.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3247893 | CR623398.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3247893 | CR624283.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3247893 | CR621827.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1748092 | NM_001007523.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1748092 | NM_001007524.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1748092 | CR623398.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1748092 | CR624283.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1748092 | CR621827.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1748092 | NM_001007523.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1748092 | NM_001007524.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1748092 | CR623398.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1748092 | CR624283.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1748092 | CR621827.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3247893 | NM_001007523.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3247893 | NM_001007524.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3247893 | CR623398.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3247893 | CR624283.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3247893 | CR621827.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3247893 | NM_001007523.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3247893 | NM_001007524.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3247893 | CR623398.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3247893 | CR624283.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3247893 | CR621827.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1748092 | NM_001007523.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1748092 | NM_001007524.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1748092 | CR623398.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1748092 | CR624283.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1748092 | CR621827.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_4937-S | - | - |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 7150161 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_4937 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1748092 | NM_001007523.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1748092 | NM_001007524.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1748092 | CR623398.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1748092 | CR624283.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1748092 | CR621827.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1727984 | NM_001007523.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1727984 | NM_001007524.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1727984 | CR623398.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1727984 | CR624283.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1727984 | CR621827.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1748092 | NM_001007523.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1748092 | NM_001007524.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1748092 | CR623398.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1748092 | CR624283.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1748092 | CR621827.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1748092 | NM_001007523.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1748092 | NM_001007524.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1748092 | CR623398.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1748092 | CR624283.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1748092 | CR621827.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1727984 | NM_001007523.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1727984 | NM_001007524.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1727984 | CR623398.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1727984 | CR624283.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1727984 | CR621827.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1727984 | NM_001007523.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1727984 | NM_001007524.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1727984 | CR623398.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1727984 | CR624283.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1727984 | CR621827.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1748092 | NM_001007523.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1748092 | NM_001007524.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1748092 | CR623398.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1748092 | CR624283.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1748092 | CR621827.1 | human_transcriptome |

F8A1
Genomic Map