Assay Details
| Gene Symbol: ICK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016513.4 | NP_057597.2 | 2 - 3 | 318 | 77 | |
| XM_011514421.1 | XP_011512723.1 | 2 - 3 | 288 | 77 | ||
| XM_017010491.1 | XP_016865980.1 | 2 - 3 | 354 | 77 | ||
| GenBank mRNA | AF225919.1 | - | 1 - 2 | 37 | 77 | |
| AK074892.1 | - | 2 - 3 | 288 | 77 | ||
| BC035807.1 | - | 1 - 2 | 239 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF225919.1, AK074892.1, BC035807.1, NM_016513.4, XM_011514421.1, XM_017010491.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
signal transduction
multicellular organism development
intracellular signal transduction
intraciliary anterograde transport
intraciliary retrograde transport
intraciliary transport
cilium assembly
regulation of cell cycle
magnesium ion binding
protein serine/threonine kinase activity
cyclin-dependent protein serine/threonine kinase activity
protein binding
ATP binding
protein phosphorylation
signal transduction
multicellular organism development
intracellular signal transduction
intraciliary anterograde transport
intraciliary retrograde transport
intraciliary transport
cilium assembly
regulation of cell cycle
magnesium ion binding
protein serine/threonine kinase activity
cyclin-dependent protein serine/threonine kinase activity
protein binding
ATP binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000350082.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | ENST00000356971.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_014920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012061.hg.1 | NM_016513.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 221886 | - | - |

ICK
Genomic Map