Assay Details
| Gene Symbol: IQCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287501.1 | NP_001274430.1 | 20 | 4488 | 87 | |
| NM_152558.4 | NP_689771.3 | 22 | 4582 | 87 | ||
| XM_017011902.1 | XP_016867391.1 | 22 | 4644 | 87 | ||
| XM_017011903.1 | XP_016867392.1 | 22 | 4522 | 87 | ||
| GenBank mRNA | AL137525.1 | - | 1 | 2303 | 87 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

IQCE
Genomic Map