| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000395878.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000395878.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000395878.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000395878.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000395878.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000395878.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000395878.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000395878.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000395878.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000395878.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000395878.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000395878.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000563366.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000563366.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000563366.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000563366.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000563366.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000563366.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000563366.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000564106.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000564106.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000564106.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000564106.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000565634.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000565634.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000565634.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000565634.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000565634.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000566136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000566136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000566136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000566136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000566136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000566669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000566669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000566669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000567212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000567212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000567212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000567212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000567212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000567212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000567212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000567212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000567212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000567212.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000567264.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000567264.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000567264.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000567264.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000569919.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000569919.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000569919.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000569919.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000569919.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000571064.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000571064.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000571064.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000571064.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000571064.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
ENST00000571064.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
NM_019116.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
NM_019116.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
NM_019116.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
NM_019116.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
NM_019116.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
NM_019116.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
NM_019116.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
NM_019116.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
NM_019116.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
NM_019116.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007229.hg.1 |
NM_019116.2 |
Refseq |