| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005909.hg.1 |
ENST00000243911.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005909.hg.1 |
NM_019888.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005909.hg.1 |
TC20000426.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007814.hg.1 |
ENST00000243911.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007814.hg.1 |
ENST00000243911.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007814.hg.1 |
NM_019888.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007814.hg.1 |
NM_019888.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007814.hg.1 |
ENST00000243911.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007814.hg.1 |
ENST00000243911.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007814.hg.1 |
NM_019888.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007814.hg.1 |
NM_019888.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000154222.hg.1 |
ENST00000243911.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154222.hg.1 |
NM_019888.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000154222.hg.1 |
TC2000007814.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000154222.hg.1 |
ENST00000243911.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154222.hg.1 |
NM_019888.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000154221.hg.1 |
ENST00000243911.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154221.hg.1 |
NM_019888.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000154221.hg.1 |
TC2000007814.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000154221.hg.1 |
ENST00000243911.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000154221.hg.1 |
NM_019888.3 |
Refseq |