Assay Details
| Gene Symbol: CCR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031200.2 | NP_112477.1 | 2 - 3 | 207 | 61 | |
| XM_011533312.2 | XP_011531614.1 | 2 - 3 | 920 | 61 | ||
| GenBank mRNA | AF145207.1 | - | 2 - 3 | 150 | 61 | |
| AF145439.1 | - | 2 - 3 | 184 | 61 | ||
| AJ132337.1 | - | 2 - 3 | 86 | 61 | ||
| AK313366.1 | - | 2 - 3 | 161 | 61 | ||
| BC069678.1 | - | 2 - 3 | 168 | 7262627 | 61 | |
| DB103756.1 | - | 2 - 3 | 207 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF145207.1, AF145439.1, AJ132337.1, AK313366.1, BC069678.1, NM_031200.2, XM_011533312.2 transcripts.
Set Membership: |
|

CCR9
Genomic Map