Assay Details
| Gene Symbol: CABP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031205.3 | NP_112482.1 | 1 - 2 | 182 | 85 | |
| XM_017020238.1 | XP_016875727.1 | 1 - 2 | 1828 | 85 | ||
| GenBank mRNA | AF169149.1 | - | 1 - 2 | 121 | 85 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000948.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12012298.hg.1 | ENST00000288616.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012298.hg.1 | ENST00000351200.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012298.hg.1 | NM_004276.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012298.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012298.hg.1 | TC12000948.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012301.hg.1 | ENST00000288616.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012301.hg.1 | ENST00000453000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012301.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12012301.hg.1 | TC12000948.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000316803.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000316803.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000316803.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000316803.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000316803.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000316803.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000316803.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000316803.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000316803.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000316803.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000351200.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000351200.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000351200.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000351200.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000351200.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000351200.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000351200.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000351200.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000453000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000453000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000453000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000453000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000453000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000453000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000453000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000453000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000453000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000498082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000498082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000498082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000498082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000498082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000498082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000498082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | ENST00000498082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_001033677.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_001033677.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_001033677.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_001033677.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_001033677.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_001033677.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_001033677.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_001033677.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_001033677.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_004276.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_004276.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_004276.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_004276.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_004276.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_004276.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_004276.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_004276.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_004276.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009136.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167559.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167559.hg.1 | ENST00000351200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167559.hg.1 | NM_004276.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167559.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167559.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167559.hg.1 | ENST00000351200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167559.hg.1 | NM_004276.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167559.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167559.hg.1 | TC1200009136.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200167562.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167562.hg.1 | ENST00000453000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167562.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167562.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167562.hg.1 | ENST00000453000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167562.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167562.hg.1 | TC1200009136.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000316803.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000316803.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000316803.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000316803.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000316803.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000316803.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000316803.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000316803.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000316803.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000316803.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000351200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000351200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000351200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000351200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000351200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000351200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000351200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000351200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000453000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000453000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000453000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000453000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000453000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000453000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000453000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000453000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000453000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000498082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000498082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000498082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000498082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000498082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000498082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000498082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | ENST00000498082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_001033677.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_001033677.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_001033677.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_001033677.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_001033677.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_001033677.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_001033677.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_001033677.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_001033677.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_004276.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_004276.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_004276.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_004276.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_004276.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_004276.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_004276.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_004276.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_004276.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009136.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167560.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167560.hg.1 | ENST00000351200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167560.hg.1 | NM_004276.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167560.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167560.hg.1 | ENST00000288616.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167560.hg.1 | ENST00000351200.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200167560.hg.1 | NM_004276.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167560.hg.1 | NM_031205.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200167560.hg.1 | TC1200009136.hg.1 | Clariom D TC |

CABP1
Genomic Map