Assay Details
| Gene Symbol: GNGT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198754.1 | NP_001185683.1 | 2 - 3 | 240 | 107 | |
| NM_001198755.1 | NP_001185684.1 | 2 - 3 | 267 | 107 | ||
| NM_001198756.1 | NP_001185685.1 | 2 - 3 | 157 | 107 | ||
| NM_031498.2 | NP_113686.1 | 2 - 3 | 284 | 107 | ||
| GenBank mRNA | AK312839.1 | - | 2 - 3 | 268 | 107 | |
| BC008663.1 | - | 2 - 3 | 284 | 3866242 | 107 | |
| BM723861.1 | - | 2 - 3 | 157 | 107 | ||
| DA248836.1 | - | 2 - 3 | 240 | 107 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001660.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK312839.1,BC008663.1,NM_001198754.1,NM_001198755.1,NM_001198756.1,NM_031498.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17022228.hg.1 | ENST00000300406.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022228.hg.1 | ENST00000503070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022228.hg.1 | ENST00000507680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022228.hg.1 | ENST00000511277.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022228.hg.1 | ENST00000511673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022228.hg.1 | ENST00000515635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022228.hg.1 | NM_001198754.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022228.hg.1 | NM_001198755.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022228.hg.1 | NM_001198756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022228.hg.1 | NM_031498.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022228.hg.1 | TC17001660.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000300406.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000300406.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000300406.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000300406.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000300406.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000300406.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000300406.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000300406.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000300406.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000300406.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000503070.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000503070.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000503070.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000503070.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000507680.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000507680.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000507680.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000507680.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000507680.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000507680.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000511277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000511277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000511277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000511277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000511277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000511277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000511277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000511277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000511277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000511673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000511673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000511673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000511673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000511673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000511673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000515635.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000515635.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000515635.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000515635.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000515635.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000515635.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | ENST00000515635.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198754.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198754.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198754.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198754.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198754.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198754.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198754.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198754.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198754.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198755.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198755.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198755.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198755.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198755.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198755.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198755.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198755.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198755.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198755.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_001198756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_031498.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_031498.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_031498.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_031498.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_031498.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_031498.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_031498.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_031498.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_031498.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_031498.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011032.hg.1 | NM_031498.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022238.hg.1 | ENST00000507680.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022238.hg.1 | NM_001198754.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17022238.hg.1 | TC17001660.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000300406.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000300406.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000300406.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000300406.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000300406.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000300406.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000300406.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000300406.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000300406.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000300406.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000503070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000503070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000503070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000503070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000507680.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000507680.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000507680.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000507680.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000507680.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000507680.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000511277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000511277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000511277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000511277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000511277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000511277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000511277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000511277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000511277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000511673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000511673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000511673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000511673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000511673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000511673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000515635.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000515635.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000515635.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000515635.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000515635.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000515635.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | ENST00000515635.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198754.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198754.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198754.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198754.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198754.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198754.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198754.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198754.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198754.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198755.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198755.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198755.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198755.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198755.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198755.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198755.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198755.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198755.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198755.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198756.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198756.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198756.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198756.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198756.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198756.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198756.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198756.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_001198756.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_031498.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_031498.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_031498.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_031498.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_031498.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_031498.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_031498.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_031498.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_031498.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_031498.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011032.hg.1 | NM_031498.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188756.hg.1 | ENST00000300406.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188756.hg.1 | ENST00000503070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188756.hg.1 | ENST00000507680.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188756.hg.1 | ENST00000511277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188756.hg.1 | ENST00000511673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188756.hg.1 | ENST00000515635.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188756.hg.1 | NM_001198754.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188756.hg.1 | NM_001198755.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188756.hg.1 | NM_001198756.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188756.hg.1 | NM_031498.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188756.hg.1 | ENST00000300406.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188756.hg.1 | ENST00000503070.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188756.hg.1 | ENST00000507680.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188756.hg.1 | ENST00000511277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188756.hg.1 | ENST00000511673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188756.hg.1 | ENST00000515635.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188756.hg.1 | NM_001198754.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188756.hg.1 | NM_001198755.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188756.hg.1 | NM_001198756.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188756.hg.1 | NM_031498.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188756.hg.1 | TC1700011032.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700188767.hg.1 | ENST00000507680.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188767.hg.1 | NM_001198754.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188767.hg.1 | ENST00000507680.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188767.hg.1 | NM_001198754.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188767.hg.1 | TC1700011032.hg.1 | Clariom D TC |

GNGT2
Genomic Map