Assay Details
| Gene Symbol: TRIM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033017.3 | NP_148977.2 | 1 - 2 | 595 | 114 | |
| GenBank mRNA | AF220023.1 | - | 1 - 2 | 532 | 114 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07003367.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000496896.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000496896.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | ENST00000496896.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025277.hg.1 | ENST00000355947.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025277.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07025277.hg.1 | TC07003367.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0700201319.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201319.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201319.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700201319.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700201319.hg.1 | TC0700013594.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000349062.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000354241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000355947.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000496896.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000496896.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | ENST00000496896.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033017.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013594.hg.1 | NM_033091.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 222729 | - | - |

TRIM4
Genomic Map