| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009454.hg.1 |
ENST00000370350.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009454.hg.1 |
NM_033085.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X009454.hg.1 |
TC0X000734.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008736.hg.1 |
ENST00000370350.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008736.hg.1 |
ENST00000370350.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008736.hg.1 |
ENST00000370350.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008736.hg.1 |
ENST00000370350.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008736.hg.1 |
ENST00000370350.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008736.hg.1 |
ENST00000370350.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008736.hg.1 |
ENST00000370350.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008736.hg.1 |
ENST00000370350.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008736.hg.1 |
ENST00000417321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008736.hg.1 |
ENST00000417321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008736.hg.1 |
ENST00000417321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008736.hg.1 |
ENST00000417321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008736.hg.1 |
ENST00000417321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008736.hg.1 |
NM_033085.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008736.hg.1 |
NM_033085.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008736.hg.1 |
NM_033085.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008736.hg.1 |
NM_033085.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008736.hg.1 |
NM_033085.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008736.hg.1 |
NM_033085.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008736.hg.1 |
NM_033085.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008736.hg.1 |
NM_033085.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008736.hg.1 |
ENST00000370350.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008736.hg.1 |
ENST00000370350.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008736.hg.1 |
ENST00000370350.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008736.hg.1 |
ENST00000370350.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008736.hg.1 |
ENST00000370350.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008736.hg.1 |
ENST00000370350.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008736.hg.1 |
ENST00000370350.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008736.hg.1 |
ENST00000370350.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008736.hg.1 |
ENST00000417321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008736.hg.1 |
ENST00000417321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008736.hg.1 |
ENST00000417321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008736.hg.1 |
ENST00000417321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008736.hg.1 |
ENST00000417321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008736.hg.1 |
NM_033085.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008736.hg.1 |
NM_033085.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008736.hg.1 |
NM_033085.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008736.hg.1 |
NM_033085.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008736.hg.1 |
NM_033085.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008736.hg.1 |
NM_033085.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008736.hg.1 |
NM_033085.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008736.hg.1 |
NM_033085.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00159368.hg.1 |
ENST00000370350.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00159368.hg.1 |
NM_033085.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00159368.hg.1 |
ENST00000370350.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00159368.hg.1 |
NM_033085.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00159368.hg.1 |
TC0X00008736.hg.1 |
Clariom D TC |