| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
ENST00000368121.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
ENST00000368121.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
ENST00000368121.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
ENST00000368121.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
ENST00000368121.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
ENST00000368122.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
ENST00000368122.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
ENST00000368122.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
ENST00000368122.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
ENST00000368122.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
ENST00000368122.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
ENST00000435307.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
ENST00000435307.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
ENST00000435307.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
NM_001122951.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
NM_001122951.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
NM_001122951.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
NM_001122951.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
NM_001122951.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018307.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021184.hg.1 |
ENST00000359381.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021184.hg.1 |
ENST00000368122.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021184.hg.1 |
ENST00000537147.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021184.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021184.hg.1 |
TC01006337.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021187.hg.1 |
NM_001122951.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021187.hg.1 |
TC01006337.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021190.hg.1 |
ENST00000359381.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021190.hg.1 |
ENST00000368121.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021190.hg.1 |
ENST00000368122.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021190.hg.1 |
ENST00000537147.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021190.hg.1 |
NM_001122951.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021190.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021190.hg.1 |
TC01006337.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021186.hg.1 |
ENST00000359381.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021186.hg.1 |
ENST00000368122.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021186.hg.1 |
ENST00000537147.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021186.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021186.hg.1 |
TC01006337.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021185.hg.1 |
ENST00000359381.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021185.hg.1 |
ENST00000368122.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021185.hg.1 |
ENST00000537147.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021185.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021185.hg.1 |
TC01006337.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021188.hg.1 |
ENST00000368121.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021188.hg.1 |
NM_001122951.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021188.hg.1 |
TC01006337.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021189.hg.1 |
ENST00000359381.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021189.hg.1 |
ENST00000368121.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021189.hg.1 |
ENST00000368122.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021189.hg.1 |
ENST00000537147.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021189.hg.1 |
NM_001122951.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021189.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021189.hg.1 |
TC01006337.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021182.hg.1 |
ENST00000359381.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021182.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021182.hg.1 |
TC01006337.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
ENST00000368121.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
ENST00000368121.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
ENST00000368121.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
ENST00000368121.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
ENST00000368121.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
ENST00000368122.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
ENST00000368122.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
ENST00000368122.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
ENST00000368122.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
ENST00000368122.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
ENST00000368122.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
ENST00000435307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
ENST00000435307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
ENST00000435307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
NM_001122951.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
NM_001122951.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
NM_001122951.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
NM_001122951.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
NM_001122951.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018307.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239763.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239763.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239763.hg.1 |
TC0100018307.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100239770.hg.1 |
ENST00000368121.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239770.hg.1 |
ENST00000435307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239770.hg.1 |
NM_001122951.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239770.hg.1 |
ENST00000368121.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239770.hg.1 |
ENST00000435307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239770.hg.1 |
NM_001122951.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239770.hg.1 |
TC0100018307.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100239766.hg.1 |
ENST00000368122.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239766.hg.1 |
ENST00000435307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239766.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239766.hg.1 |
ENST00000368122.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239766.hg.1 |
ENST00000435307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239766.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239766.hg.1 |
TC0100018307.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100239765.hg.1 |
ENST00000368122.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239765.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239765.hg.1 |
ENST00000368122.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239765.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239765.hg.1 |
TC0100018307.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100239771.hg.1 |
ENST00000368121.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239771.hg.1 |
ENST00000368122.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239771.hg.1 |
ENST00000435307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239771.hg.1 |
NM_001122951.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239771.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239771.hg.1 |
ENST00000368121.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239771.hg.1 |
ENST00000368122.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239771.hg.1 |
ENST00000435307.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239771.hg.1 |
NM_001122951.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239771.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239771.hg.1 |
TC0100018307.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100239764.hg.1 |
ENST00000368122.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239764.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239764.hg.1 |
ENST00000368122.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239764.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239764.hg.1 |
TC0100018307.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100239768.hg.1 |
ENST00000368121.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239768.hg.1 |
NM_001122951.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239768.hg.1 |
ENST00000368121.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239768.hg.1 |
NM_001122951.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239768.hg.1 |
TC0100018307.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100239772.hg.1 |
ENST00000368121.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239772.hg.1 |
ENST00000368122.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239772.hg.1 |
NM_001122951.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239772.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239772.hg.1 |
ENST00000368121.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239772.hg.1 |
ENST00000368122.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239772.hg.1 |
NM_001122951.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239772.hg.1 |
NM_002036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239772.hg.1 |
TC0100018307.hg.1 |
Clariom D TC |