| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009723.hg.1 |
ENST00000440592.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050026.hg.1 |
ENST00000271638.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050026.hg.1 |
ENST00000478109.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050026.hg.1 |
NM_005620.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050026.hg.1 |
TC01003240.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006628.hg.1 |
ENST00000487308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009605.hg.1 |
ENST00000603331.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007987.hg.1 |
ENST00000545814.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007232.hg.1 |
ENST00000527476.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007299.hg.1 |
ENST00000599675.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700008664.hg.1 |
ENST00000477525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050027.hg.1 |
ENST00000271638.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050027.hg.1 |
NM_005620.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050027.hg.1 |
TC01003240.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007169.hg.1 |
ENST00000467589.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015822.hg.1 |
ENST00000271638.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015822.hg.1 |
ENST00000271638.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015822.hg.1 |
ENST00000271638.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015822.hg.1 |
ENST00000271638.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015822.hg.1 |
ENST00000271638.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015822.hg.1 |
ENST00000478109.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015822.hg.1 |
ENST00000478109.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015822.hg.1 |
ENST00000478109.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015822.hg.1 |
ENST00000478109.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015822.hg.1 |
NM_005620.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015822.hg.1 |
NM_005620.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015822.hg.1 |
NM_005620.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015822.hg.1 |
NM_005620.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015822.hg.1 |
NM_005620.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050029.hg.1 |
ENST00000271638.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050029.hg.1 |
NM_005620.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050029.hg.1 |
TC01003240.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0700006628.hg.1 |
ENST00000487308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009723.hg.1 |
ENST00000440592.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007987.hg.1 |
ENST00000545814.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015822.hg.1 |
ENST00000271638.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015822.hg.1 |
ENST00000271638.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015822.hg.1 |
ENST00000271638.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015822.hg.1 |
ENST00000271638.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015822.hg.1 |
ENST00000271638.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015822.hg.1 |
ENST00000478109.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015822.hg.1 |
ENST00000478109.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015822.hg.1 |
ENST00000478109.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015822.hg.1 |
ENST00000478109.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015822.hg.1 |
NM_005620.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015822.hg.1 |
NM_005620.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015822.hg.1 |
NM_005620.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015822.hg.1 |
NM_005620.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015822.hg.1 |
NM_005620.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700008664.hg.1 |
ENST00000477525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007232.hg.1 |
ENST00000527476.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007169.hg.1 |
ENST00000467589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218883.hg.1 |
ENST00000271638.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218883.hg.1 |
NM_005620.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100218883.hg.1 |
ENST00000271638.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218883.hg.1 |
NM_005620.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100218883.hg.1 |
TC0100015822.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00009605.hg.1 |
ENST00000603331.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007299.hg.1 |
ENST00000599675.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218879.hg.1 |
ENST00000271638.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218879.hg.1 |
ENST00000478109.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218879.hg.1 |
NM_005620.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100218879.hg.1 |
ENST00000271638.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218879.hg.1 |
ENST00000478109.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218879.hg.1 |
NM_005620.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100218879.hg.1 |
TC0100015822.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100218881.hg.1 |
ENST00000271638.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218881.hg.1 |
NM_005620.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100218881.hg.1 |
ENST00000271638.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218881.hg.1 |
NM_005620.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100218881.hg.1 |
TC0100015822.hg.1 |
Clariom D TC |