Assay Details
| Gene Symbol: RARB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC069735.1 | - | 1 | 450 | 7262624 | 146 |
| M62303.1 | - | 1 | 456 | 149 | ||
| S82362.1 | - | 1 | 1443 | 146 | ||
More Information
Updated Mapping Information
Set Membership: |
|

RARB
Genomic Map