| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05001536.hg.1 |
ENST00000330120.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05001536.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05001536.hg.1 |
TC05000149.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007097.hg.1 |
ENST00000330120.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007097.hg.1 |
ENST00000330120.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007097.hg.1 |
ENST00000330120.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007097.hg.1 |
ENST00000330120.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007097.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007097.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007097.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007097.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007097.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05001538.hg.1 |
ENST00000330120.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05001538.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05001538.hg.1 |
TC05000149.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0500007097.hg.1 |
ENST00000330120.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007097.hg.1 |
ENST00000330120.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007097.hg.1 |
ENST00000330120.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007097.hg.1 |
ENST00000330120.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007097.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007097.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007097.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007097.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007097.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500147908.hg.1 |
ENST00000330120.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500147908.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500147908.hg.1 |
ENST00000330120.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500147908.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500147908.hg.1 |
TC0500007097.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500147910.hg.1 |
ENST00000330120.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500147910.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500147910.hg.1 |
ENST00000330120.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500147910.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500147910.hg.1 |
TC0500007097.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500147911.hg.1 |
ENST00000330120.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500147911.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500147911.hg.1 |
ENST00000330120.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500147911.hg.1 |
NM_016568.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500147911.hg.1 |
TC0500007097.hg.1 |
Clariom D TC |