Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PPP2R2B intronic transcript 1 | PPP2R2B-IT1 | -19.3kb | Sense | Chr. 5 - 146914207 - 146919506 on NCBI Build GRCh38 | |
| Downstream | serine/threonine kinase 32A | STK32A | YANK1 | 294.3kb | Anti-sense | Chr. 5 - 147234963 - 147401996 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map