Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 25 member 19 | SLC25A19 |
DNC, MCPHA, MUP1, THMD3, THMD4, TPC |
-81.0kb | Sense | Chr. 17 - 75272980 - 75289957 on NCBI Build GRCh38 |
| Downstream | microRNA 3678 | MIR3678 | mir-3678 | 32.3kb | Anti-sense | Chr. 17 - 75406069 - 75406162 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map