Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928847 | LOC101928847 | -304.4kb | Sense | Chr. 11 - 112959279 - 112963460 on NCBI Build GRCh38 | |
| Downstream | tetratricopeptide repeat domain 12 | TTC12 | TPARM | 44.8kb | Anti-sense | Chr. 11 - 113314529 - 113385952 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map