| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009985.hg.1 |
ENST00000628401.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009985.hg.1 |
NR_103844.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009985.hg.1 |
NR_103844.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009985.hg.1 |
NR_103844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500174306.hg.1 |
NR_103844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500174306.hg.1 |
TC1500009985.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500174306.hg.1 |
NR_103844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500174307.hg.1 |
ENST00000628401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500174307.hg.1 |
NR_103844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500174307.hg.1 |
TC1500009985.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500174307.hg.1 |
ENST00000628401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500174307.hg.1 |
NR_103844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009985.hg.1 |
ENST00000628401.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009985.hg.1 |
NR_103844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009985.hg.1 |
NR_103844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009985.hg.1 |
NR_103844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500174308.hg.1 |
NR_103844.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500174308.hg.1 |
TC1500009985.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500174308.hg.1 |
NR_103844.1 |
Refseq |