Assay Details
| Gene Symbol: LOC219690 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK055023.1 | - | 1 - 2 | 46 | 69 | |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686901 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16353234 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304147 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304123 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686971 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304117 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27687022 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304212 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304187 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304113 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686915 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686877 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304234 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27687017 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304179 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686999 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16353218 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304184 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304119 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686940 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304148 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304228 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304154 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304239 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304112 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27687030 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16353227 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304190 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686943 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304174 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304182 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304183 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304150 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304205 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304151 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304114 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304213 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304230 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304163 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304207 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27687021 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304191 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304134 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27687024 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16353212 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304186 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304102 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304218 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304255 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304202 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304223 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686998 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304103 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304137 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304146 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686875 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304160 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304198 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304125 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686951 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304162 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304211 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686991 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686883 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304159 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686944 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16353232 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686973 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304185 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686902 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304220 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16353236 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686974 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304158 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304192 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304203 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304206 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304222 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304188 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304249 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304166 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304169 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304121 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686881 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304208 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686886 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686976 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27687025 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686934 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686990 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304189 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304139 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304127 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304152 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686907 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304178 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686946 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304236 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304132 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304215 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304251 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304104 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304232 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686912 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686925 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304193 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304105 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304138 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304248 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304201 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686905 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304129 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304165 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686913 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27687020 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304180 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304126 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304140 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304226 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16353237 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304153 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27686931 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07304136 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27687023 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 566341 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 920448 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3508160 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 28545 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3508160 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18970297 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P18970297 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1564314_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007941.hg.1 | ENST00000603033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007941.hg.1 | ENST00000603033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007941.hg.1 | ENST00000603033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007941.hg.1 | ENST00000603033.1 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 1564314_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1564314_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564314_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564314_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564314_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564314_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564314_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564314_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564314_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564314_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564314_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564314_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564314_at5 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina human-6 v2.0 expression beadchip | ILMN_1827895 | - | - |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 2850544 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_91107 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1827895 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1827895 | - | - |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1827895 | - | - |

LOC219690
Genomic Map