Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | eukaryotic translation initiation factor 3 subunit J pseudogene | LOC100422212 | -1.2mb | Anti-sense | Chr. 1 - 163421070 - 163423191 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC100505795 | LOC100505795 | 166.4kb | Anti-sense | Chr. 1 - 164769116 - 164774641 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map