Assay Details
| Gene Symbol: ELFN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK055475.1 | - | 1 - 2 | 435 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100506271 | LOC100506271 | Sense | Chr. 22 - 37352190 - 37354839 on NCBI Build GRCh38 | ||
| Downstream | MFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase | MFNG | 83.8kb | Sense | Chr. 22 - 37469063 - 37486440 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000415408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000415408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000415408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000415408.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000435824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000435824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000435824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000435824.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000451509.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000451509.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000451509.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000452946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000452946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000452946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NR_110512.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NR_110512.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NR_110512.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008643.hg.1 | NR_110512.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000402918.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000414347.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000415408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000415408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000415408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000415408.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000424973.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000430883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000435824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000435824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000435824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000435824.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000451509.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000451509.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000451509.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000452946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000452946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000452946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | ENST00000613079.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NM_052906.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NR_110512.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NR_110512.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NR_110512.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008643.hg.1 | NR_110512.1 | Refseq |

ELFN2
Genomic Map