Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Rho GTPase activating protein 24 | ARHGAP24 |
FILGAP, RC-GAP72, RCGAP72, p73, p73RhoGAP |
Sense | Chr. 4 - 85475131 - 86002670 on NCBI Build GRCh38 | |
| Downstream | microRNA 4451 | MIR4451 | mir-4451 | 44.4kb | Sense | Chr. 4 - 85722468 - 85722533 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map