Assay Details
| Gene Symbol: WDFY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014991.4 | NP_055806.2 | 51 - 52 | 8450 | 72 | |
| XM_005262858.4 | XP_005262915.1 | 53 - 54 | 8491 | 72 | ||
| XM_011531757.2 | XP_011530059.1 | 52 - 53 | 8402 | 72 | ||
| XM_011531759.2 | XP_011530061.1 | 51 - 52 | 8364 | 72 | ||
| XM_011531760.2 | XP_011530062.1 | 51 - 52 | 8302 | 72 | ||
| XM_011531761.2 | XP_011530063.1 | 52 - 53 | 8357 | 72 | ||
| XM_011531762.2 | XP_011530064.1 | 51 - 52 | 8351 | 72 | ||
| XM_011531763.2 | XP_011530065.1 | 50 - 51 | 8248 | 72 | ||
| XM_011531764.2 | XP_011530066.1 | 51 - 52 | 8318 | 72 | ||
| XM_011531765.2 | XP_011530067.1 | 48 - 49 | 8053 | 72 | ||
| XM_017007906.1 | XP_016863395.1 | 54 - 55 | 8619 | 72 | ||
| XM_017007907.1 | XP_016863396.1 | 54 - 55 | 8616 | 72 | ||
| XM_017007908.1 | XP_016863397.1 | 53 - 54 | 8548 | 72 | ||
| XM_017007909.1 | XP_016863398.1 | 39 - 40 | 6244 | 72 | ||
| GenBank mRNA | AB023210.2 | - | 16 - 17 | 2133 | 72 | |
| AF538685.1 | - | 50 - 51 | 8342 | 72 | ||
| AK055806.1 | - | 2 - 3 | 97 | 72 | ||
| AK094910.1 | - | 7 - 8 | 851 | 72 | ||
| BC150246.1 | - | 16 - 17 | 2154 | 72 | ||
| BX648077.1 | - | 8 - 9 | 1015 | 72 | ||
| BX648153.1 | - | 11 - 12 | 1343 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001355.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK055806.1 transcripts.
Set Membership: |
|

WDFY3
Genomic Map