Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | GDNF family receptor alpha like | GFRAL |
C6orf144, GRAL, UNQ9356, bA360D14.1 |
-42.9kb | Anti-sense | Chr. 6 - 55327469 - 55402493 on NCBI Build GRCh38 |
| Downstream | bone morphogenetic protein 5 | BMP5 | 305.7kb | Sense | Chr. 6 - 55753653 - 55875625 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map