Assay Details
| Gene Symbol: LINC00628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027022.1 | 1 | 648 | 109 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003731.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00741826 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20803184 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00741834 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20803196 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20803178 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20803202 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00741828 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00741837 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00741825 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00741815 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20803199 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00741812 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00741804 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00741817 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00741821 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20803217 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20803186 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15165322 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00741831 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20803195 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00741830 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00741829 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20803194 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00741806 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00741832 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00741810 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20803180 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00741818 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00741809 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00741827 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00741833 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00741840 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00741808 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 410822 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3270172 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4939 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4938 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 6924 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3270172 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3270172 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741818 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741809 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741830 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741832 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741821 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741825 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20803202 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741834 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20803192 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741815 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20803169 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20803187 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741795 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15165308 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15165309 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741808 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20803164 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20803186 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741804 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741812 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20803194 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741799 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741810 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15165307 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741829 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20803195 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741816 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741837 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20803199 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741838 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20803180 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741833 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20803186 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20803202 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15165307 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741810 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741821 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15165309 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741795 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741838 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20803194 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741832 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20803187 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741837 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741815 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741812 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20803164 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741816 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741830 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741833 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15165308 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20803199 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741799 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20803192 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20803180 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741809 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741825 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741818 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20803169 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741834 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20803195 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741804 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741829 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00741808 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P749494 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741762 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741840 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15165312 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15165313 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741732 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741800 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741804 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741837 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741758 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15165309 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741773 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741734 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741755 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741794 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741818 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741832 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741810 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741742 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741739 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741823 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741775 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741737 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741767 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741833 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15165322 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741752 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741802 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741769 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741748 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741751 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741834 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741733 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741824 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741809 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741820 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741807 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741744 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741821 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741736 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15165316 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741763 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741816 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741771 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741830 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15165302 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741813 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741801 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741765 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741814 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741829 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00741798 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1565556_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1565554_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057276.hg.1 | NR_027022.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057276.hg.1 | TC01003731.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017026.hg.1 | ENST00000628516.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017026.hg.1 | ENST00000628516.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017026.hg.1 | NR_027022.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017026.hg.1 | NR_027022.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017026.hg.1 | ENST00000628516.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017026.hg.1 | ENST00000628516.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017026.hg.1 | NR_027022.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017026.hg.1 | NR_027022.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229016.hg.1 | ENST00000628516.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229016.hg.1 | NR_027022.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229016.hg.1 | ENST00000628516.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229016.hg.1 | NR_027022.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229016.hg.1 | TC0100017026.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100229015.hg.1 | ENST00000628516.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229015.hg.1 | NR_027022.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229015.hg.1 | ENST00000628516.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229015.hg.1 | NR_027022.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229015.hg.1 | TC0100017026.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1565556_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1565554_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1565556_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1565554_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565554_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565556_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565554_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565554_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565556_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565556_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565556_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565554_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565554_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565556_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565556_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565554_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565556_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565554_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565554_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565556_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565554_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565556_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565554_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565554_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565556_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565556_at3 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A010086 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A010086 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1886412 | NR_027022.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1886412 | BC015457.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1886412 | NR_027022.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1886412 | BC015457.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1886412 | NR_027022.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1886412 | BC015457.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_82354 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_82354 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1886412 | NR_027022.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1886412 | BC015457.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1886412 | NR_027022.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1886412 | BC015457.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1886412 | NR_027022.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1886412 | BC015457.1 | human_transcriptome |

LINC00628
Genomic Map