Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | metastasis suppressor 1 | MTSS1 |
MIM, MIMA, MIMB |
Sense | Chr. 8 - 124550770 - 124728507 on NCBI Build GRCh38 | |
| Downstream | microRNA 4662b | MIR4662B | 207.3kb | Sense | Chr. 8 - 124821978 - 124822058 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map