| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200011918.hg.1 |
ENST00000602445.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006900.hg.1 |
ENST00000272224.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006900.hg.1 |
ENST00000272224.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006900.hg.1 |
ENST00000272224.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006900.hg.1 |
ENST00000272224.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006900.hg.1 |
ENST00000272224.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006900.hg.1 |
ENST00000272224.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006900.hg.1 |
ENST00000565841.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006900.hg.1 |
NM_182828.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006900.hg.1 |
NM_182828.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006900.hg.1 |
NM_182828.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200006900.hg.1 |
NM_182828.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200011918.hg.1 |
ENST00000602445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006900.hg.1 |
ENST00000272224.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006900.hg.1 |
ENST00000272224.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006900.hg.1 |
ENST00000272224.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006900.hg.1 |
ENST00000272224.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006900.hg.1 |
ENST00000272224.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006900.hg.1 |
ENST00000272224.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006900.hg.1 |
ENST00000565841.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200006900.hg.1 |
NM_182828.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006900.hg.1 |
NM_182828.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006900.hg.1 |
NM_182828.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200006900.hg.1 |
NM_182828.3 |
Refseq |