| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010873.hg.1 |
ENST00000450995.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010873.hg.1 |
ENST00000450995.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010873.hg.1 |
ENST00000450995.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010873.hg.1 |
ENST00000450995.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010873.hg.1 |
ENST00000450995.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010873.hg.1 |
NR_120648.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010873.hg.1 |
NR_120648.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010873.hg.1 |
NR_120648.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010873.hg.1 |
NR_120648.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010873.hg.1 |
NR_120648.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010873.hg.1 |
ENST00000450995.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010873.hg.1 |
ENST00000450995.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010873.hg.1 |
ENST00000450995.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010873.hg.1 |
ENST00000450995.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010873.hg.1 |
ENST00000450995.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010873.hg.1 |
NR_120648.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010873.hg.1 |
NR_120648.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010873.hg.1 |
NR_120648.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010873.hg.1 |
NR_120648.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010873.hg.1 |
NR_120648.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000174100.hg.1 |
ENST00000450995.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000174100.hg.1 |
NR_120648.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000174100.hg.1 |
ENST00000450995.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000174100.hg.1 |
NR_120648.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000174100.hg.1 |
TC1000010873.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000174101.hg.1 |
ENST00000450995.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000174101.hg.1 |
NR_120648.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000174101.hg.1 |
ENST00000450995.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000174101.hg.1 |
NR_120648.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000174101.hg.1 |
TC1000010873.hg.1 |
Clariom D TC |