Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | IPO9 antisense RNA 1 | IPO9-AS1 | Sense | Chr. 1 - 201688256 - 201829559 on NCBI Build GRCh38 | ||
| Downstream | microRNA 1231 | MIR1231 |
MIRN1231, hsa-mir-1231 |
71.1kb | Anti-sense | Chr. 1 - 201808611 - 201808702 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map