Assay Details
| Gene Symbol: N4BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153029.3 | NP_694574.3 | 7 | 4326 | 106 | |
| XM_011523482.1 | XP_011521784.1 | 6 | 4223 | 106 | ||
| GenBank mRNA | AK056383.1 | - | 1 | 349 | 106 | |
| AK124782.1 | - | 1 | 1012 | 106 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

N4BP1
Genomic Map