Assay Details
| Gene Symbol: LOC101927410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110777.1 | 1 - 2 | 243 | 65 | ||
| GenBank mRNA | AK056572.1 | - | 1 - 2 | 243 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NSF attachment protein gamma | NAPG | GAMMASNAP | -109.2kb | Sense | Chr. 18 - 10525876 - 10552769 on NCBI Build GRCh38 |
| Downstream | piezo type mechanosensitive ion channel component 2 | PIEZO2 |
C18orf30, C18orf58, DA3, DA5, FAM38B, FAM38B2, HsT748, HsT771, MWKS |
6.6kb | Anti-sense | Chr. 18 - 10670237 - 11149561 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101927410
Genomic Map