Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neuron navigator 2 | NAV2 |
HELAD1, POMFIL2, RAINB1, STEERIN2, UNC53H2 |
-12.0kb | Sense | Chr. 11 - 19345200 - 20121601 on NCBI Build GRCh38 |
| Downstream | developing brain homeobox 1 | DBX1 | 20.7kb | Anti-sense | Chr. 11 - 20156214 - 20160324 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map