Assay Details
| Gene Symbol: STARD7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046322.1 | 2 - 3 | 756 | 111 | ||
| GenBank mRNA | AK057419.1 | - | 2 - 3 | 228 | 111 | |
| AK311432.1 | - | 2 - 3 | 771 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01342921 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01342901 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01342908 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01342911 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01342893 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21442742 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21442721 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01342887 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01342902 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01342907 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01342895 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21442731 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01342913 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01342903 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01342909 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01342891 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01342904 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21442719 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01342896 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01342897 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01342886 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21442732 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21442724 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01342915 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15285907 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21442723 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 157612 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 513373 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 142718 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3376541 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1223 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_1222 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P687131 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 56160 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P00425759 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3376541 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3376541 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342893 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342890 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21442719 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21442721 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342909 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342907 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342915 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342911 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21442724 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342901 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342902 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21442723 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21442732 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21442731 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342908 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21442742 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21442718 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342887 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342892 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342895 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342904 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342892 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21442719 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342901 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21442731 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342887 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342909 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342908 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21442732 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342915 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342907 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21442723 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21442721 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21442718 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342895 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21442724 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342904 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342911 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342890 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21442742 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342893 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01342902 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P687131 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P00425759 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P00425759 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 53416 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 53769 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00425759 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P322865 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01342891 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01342908 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01342918 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01342885 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01342914 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01342898 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01342900 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01342904 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01342893 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01342887 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01342895 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01342902 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01342890 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01342917 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01342911 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01342919 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00425759 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_001037228_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8043584 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8043583 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 235584_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11735232_a_at | - | - |
| Affymetrix Human Genome U219 Array | 11759154_a_at | - | - |
| Affymetrix Human Genome U219 Array | 11735232_PM_a_at | - | - |
| Affymetrix Human Genome U219 Array | 11759154_PM_a_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008499.hg.1 | ENST00000432267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008499.hg.1 | ENST00000432267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008499.hg.1 | ENST00000432267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008499.hg.1 | ENST00000432267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008499.hg.1 | ENST00000432267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008499.hg.1 | ENST00000432267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008499.hg.1 | ENST00000446816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008499.hg.1 | ENST00000446816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008499.hg.1 | ENST00000446816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008499.hg.1 | ENST00000446816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008499.hg.1 | ENST00000614453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008499.hg.1 | ENST00000614453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008499.hg.1 | ENST00000614453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008499.hg.1 | NR_046322.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008499.hg.1 | NR_046322.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008499.hg.1 | NR_046322.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008499.hg.1 | NR_046322.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008499.hg.1 | NR_046322.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008499.hg.1 | NR_046322.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008499.hg.1 | NR_046322.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008499.hg.1 | NR_046322.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158935.hg.1 | NR_046322.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158935.hg.1 | NR_046322.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158935.hg.1 | TC0200008499.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200158937.hg.1 | ENST00000432267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158937.hg.1 | NR_046322.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158937.hg.1 | ENST00000432267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158937.hg.1 | NR_046322.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158937.hg.1 | TC0200008499.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200158934.hg.1 | ENST00000446816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158934.hg.1 | NR_046322.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158934.hg.1 | ENST00000446816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200158934.hg.1 | NR_046322.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200158934.hg.1 | TC0200008499.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200008499.hg.1 | ENST00000432267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008499.hg.1 | ENST00000432267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008499.hg.1 | ENST00000432267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008499.hg.1 | ENST00000432267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008499.hg.1 | ENST00000432267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008499.hg.1 | ENST00000432267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008499.hg.1 | ENST00000446816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008499.hg.1 | ENST00000446816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008499.hg.1 | ENST00000446816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008499.hg.1 | ENST00000446816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008499.hg.1 | ENST00000614453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008499.hg.1 | ENST00000614453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008499.hg.1 | ENST00000614453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008499.hg.1 | NR_046322.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008499.hg.1 | NR_046322.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008499.hg.1 | NR_046322.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008499.hg.1 | NR_046322.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008499.hg.1 | NR_046322.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008499.hg.1 | NR_046322.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008499.hg.1 | NR_046322.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008499.hg.1 | NR_046322.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 235584_at | - | - |
| Human Genome U133 Plus 2.0 Array | 235584_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235584_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235584_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235584_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235584_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235584_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235584_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235584_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235584_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235584_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235584_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235584_at11 | - | - |
| Human Genome U95 Set | 51869_at | - | - |
| Human Genome U95 Set | 51869_PM_at | - | - |
| Human Genome U95 Set | 68930_r_at | - | - |
| Human Genome U95 Set | 68930_PM_r_at | - | - |
| Human Genome U133 Set | 235584_PM_at | - | - |
| Human Genome U133 Set | 235584_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A016007 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A016007 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 232187 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_2115453 | BC101730.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2115453 | BC101732.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2115453 | CR619653.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2115453 | NM_001037228.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2115453 | AK057419.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1740601 | - | - |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1740601 | - | - |
| HumanHT-12 V4.0 expression beadchip | ILMN_2115453 | BC101730.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2115453 | BC101732.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2115453 | CR619653.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2115453 | NM_001037228.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2115453 | AK057419.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2115453 | BC101730.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2115453 | BC101732.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2115453 | CR619653.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2115453 | NM_001037228.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2115453 | AK057419.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2115453 | BC101730.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2115453 | BC101732.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2115453 | CR619653.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2115453 | NM_001037228.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2115453 | AK057419.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1740601 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2115453 | BC101730.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2115453 | BC101732.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2115453 | CR619653.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2115453 | NM_001037228.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2115453 | AK057419.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2115453 | BC101730.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2115453 | BC101732.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2115453 | CR619653.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2115453 | NM_001037228.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2115453 | AK057419.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1740601 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2115453 | BC101730.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2115453 | BC101732.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2115453 | CR619653.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2115453 | NM_001037228.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2115453 | AK057419.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1740601 | - | - |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2115453 | BC101730.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2115453 | BC101732.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2115453 | CR619653.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2115453 | NM_001037228.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2115453 | AK057419.1 | human_transcriptome |

STARD7-AS1
Genomic Map